Description
Eliminate comb assembly with Thermo Scientific™ Owl™ S3S System Combs. These heavy-duty, one-piece combs are for use with the Owl™ S3S System.
Specifications
Specifications
| Description | Sharktooth Comb |
| For Use With (Equipment) | Owl™ S3S Aluminum-Backed Sequencer System |
| Product Type | Combs |
| Teeth Count | 110 (3X) |
| Thickness (Metric) Teeth | 0.4 mm |
| Volume (Metric) Well | 5.5µL |