Learn More
Thermo Scientific™ Owl™ S3S System Combs
Catalog No. OWS2SMT44
Change view
Click to view available options
Description:
Sharktooth Comb
Sharktooth Micro-Well Comb
Teeth Count:
110 (3X)
146 (4X)
72 (2X)
Thickness (Metric) Teeth:
0.4 mm
Volume (Metric) Well:
10µL
3.7µL
5.5µL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Teeth Count Thickness (Metric) Teeth Volume (Metric) Well
OW-S2S-MT44 Sharktooth Micro-Well Comb 146 (4X) 0.4 mm 3.7µL
OW-S2S-MT2 Sharktooth Micro-Well Comb 72 (2X) 0.4 mm 10µL
OW-S2S-MT3 Sharktooth Comb 110 (3X) 0.4 mm 5.5µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. OW-S2S-MT44 Supplier Thermo Scientific™ Supplier No. S2SMT44
Add to Cart
Add to Cart
Eliminate comb assembly with these heavy-duty, one-piece combs.
Eliminate comb assembly with Thermo Scientific™ Owl™ S3S System Combs. These heavy-duty, one-piece combs are for use with the Owl™ S3S System.
Heavy duty, one piece combs eliminate comb assembly.
TRUSTED_SUSTAINABILITY
Specifications
Description Sharktooth Micro-Well Comb
For Use With (Equipment) Owl™ S3S Aluminum-Backed Sequencer System
Product Type Combs
Teeth Count 146 (4X)
Thickness (Metric) Teeth 0.4 mm
Volume (Metric) Well 3.7µL