Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Owl™ VEP-2 Mini Tank Electroblotting System
SCI_ED

Catalog No. S35837 Shop All Thermo Scientific Products
Change view
Click to view available options
Dimensions (L x W x H):
18 x 13 x 16.5 cm
Capacity:
Transfer Area: 9 x 9 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H) Capacity
S35837 18 x 13 x 16.5 cm Transfer Area: 9 x 9 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S35837 Supplier Thermo Scientific™ Supplier No. VEP2/EMD

missing translation for 'validMainframeMsg'

Produce uniform, reproducible protein transfers for up to four polyacrylamide minigels with the Thermo Scientific™ Owl™ VEP-2 Mini Tank Electroblotting System.

Features:
  • Tank-style electroblotting system
  • Integrated cooling base enables temperature-controlled runs
  • Stir bars (not included) may be added to bottom of buffer chamber for increased buffer circulation

Includes:

  • Buffer chamber with platinum wire electrode panels and cooling base
  • Safety interlocking lid with attached power supply leads
  • Four blotting cassettes with foam pads and cassette clips
  • Six sheets of blotting filter paper
TRUSTED_SUSTAINABILITY

Specifications

Capacity Transfer Area: 9 x 9 cm
Dimensions (L x W x H) 18 x 13 x 16.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.