Learn More
Athens Research And Technology Inc OX-LDL LIPOPROTEINS
Supplier:  Athens Research And Technology Inc 1216120412OX1MG
Encompass_Preferred
Lipoproteins, Oxidized LDL (OX-LDL), Human Plasma. MW 2,300,000. Liquid in PBS +0.2mM EDTA, pH 7.4. Density: 1.02 to 1.063 g/ml; Composition: 78-80% lipid, 22-20% protein. Shelf life: *6 weeks. Storage: 2-6 Degrees C; Do NOT Freeze; Store Away from Light. 1mg.
Catalog No. NC0955945
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.