Learn More
ACROBiosystems Recombinant Protein;Human OX40 Ligand / TNFSF4 Protein (active trimer) (HPLC-verified);HEK293;1MG;OXL,OX40L,TNFSF4,CD252,Glycoprotein Gp34,TXGP1
Supplier:  ACROBiosystems OXLH52Q81MG
Encompass_Preferred
Recombinant Protein;Human OX40 Ligand / TNFSF4 Protein (active trimer) (HPLC-verified);1MG;Human OX40 Ligand (active trimer) (HPLC-verified) (OXL-H52Q8) is expressed from human 293 cells (HEK293). It contains AA Gln 51 - Leu 183 (Accession # NP_003317).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 16.9 kDa. The protein migrates as 25-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human OX40, Fc Tag (Cat. No. OX0-H5255) at 2 μg/mL (100 μL/well) can bind Human OX40 Ligand (active trimer) (HPLC-verified) (Cat. No. OXL-H52Q8) with a linear range of 2-31 ng/mL (QC tested).;FACS assay shows that Human OX40 Ligand (active trimer) (HPLC-verified) (Cat. No. OXL-H52Q8) can bind to 293T cell overexpressing human OX40. The concentration of OX40 Ligand used is 0.1 μg/ml (Routinely tested).
Catalog No. 50-165-8888