Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ OXA (17-33)
GREENER_CHOICESDP

Catalog No. 51151 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-151 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 51-151 Supplier Tocris Bioscience™ Supplier No. 5115/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective peptide OX1 agonist

Potent and selective peptide orexin OX1 receptor agonist (EC50 values are 8.29 and 187 nM for OX1 and OX2 receptors respectively). Truncated form of orexin A (Cat. No. 1455).

Chemical Identifiers

CAS 343268-91-7

Specifications

Content And Storage Store at -20°C
Molecular Weight (g/mol) 1749.008
Product Type OXA (17-33)
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.