Learn More
Oxalacetic acid, 97%
Description
Oxalacetic acid is used as a metabolic intermediate in many processes that occur in animals and citric acid cycle. It is a used to converted to aspartic acid by aspartate transaminase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxalacetic acid is used as a metabolic intermediate in many processes that occur in animals and citric acid cycle. It is a used to converted to aspartic acid by aspartate transaminase.
Solubility
Soluble in water (0.1 g/ml), ether, ethanol, acetone, and acetone ethyl acetate.
Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from heat and sunlight.
Specifications
Specifications
| Melting Point | 161°C (decomposition) |
| Flash Point | 88°C (190°F) |
| Quantity | 25 g |
| UN Number | UN3261 |
| Beilstein | 1705475 |
| Merck Index | 14,6909 |
| Solubility Information | Soluble in water (0.1g/ml),ether,ethanol,acetone,and acetone ethyl acetate. |
| Formula Weight | 132.07 |
| Percent Purity | 97% |
| Chemical Name or Material | Oxalacetic acid |
Safety and Handling
| Hazard Category | H314 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
| EINECSNumber | 206-329-8 |
| TSCA | Yes |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.