Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalic Acid, 0.100 Normal (N/10), Reagents

Catalog No. 01334569
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-569 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-569 Supplier Reagents Holdings Llc Supplier No. CO1000001C
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxalic Acid, 0.100 Normal (N/10), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 6153-56-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Ethanedioic Acid
IUPAC Name water
SMILES O

Specifications

pH <2
Boiling Point 100°C
Color Colorless
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1.0025
Grade Reagent
Chemical Name or Material Oxalic Acid, 0.100 Normal (N/10)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.