Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalic Acid, 7.5% (w/v), Ricca Chemical
SDP

Catalog No. R546700020F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R546700020F 20 L Cubitainer
5467-16 500 mL Natural Poly Bottle
5467-32 1 L Natural Poly Bottle
5467-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R546700020F Supplier Ricca Chemical Company Supplier No. 54675
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 6153-56-6
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Color Colorless
Solubility Information Solubility in Water: Infinite
Percent Purity 7.5% w/v
Grade Laboratory
Quantity 20 L
Odor Odorless
Packaging Cubitainer
Physical Form Liquid
Chemical Name or Material Oxalic Acid, 7.5% (w/v), 0.4 Molar (0.8 N)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.