Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalic Acid Dihydrate, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCO0200125 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
125 g
500 g
Packaging:
Poly Wide Mouth Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCO0200125 125 g Poly Wide Mouth Bottle
RDCO0200500 500 g Poly Wide Mouth Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RDCO0200125 Supplier Ricca Chemical Company Supplier No. RDCO0200125B1
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6153-56-6
Molecular Formula C2H6O6
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N

Specifications

CAS Max % 102.5
SMILES O.O.OC(=O)C(O)=O
IUPAC Name oxalic acid dihydrate
Molecular Weight (g/mol) 126.06
Percent Purity 99.5 to 102.5%
Grade ACS
Quantity 125 g
Packaging Poly Wide Mouth Bottle
Residue after Ignition max. 0.01%
Chemical Name or Material Oxalic Acid
ShelfLife 84 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.