Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalic Acid Dihydrate, MP Biomedicals™

Catalog No. 0215255280 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-152-55280 100 g
ICN15255290 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-152-55280 Supplier MP Biomedicals, Inc Supplier No. 0215255280
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6153-56-6
Molecular Formula C2H6O6
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N

Specifications

Melting Point 101°C to 102°C
Boiling Point Sublimates
Solubility Information 13 to 14 g/100 mL at 20°C (in water)
SMILES O.O.OC(=O)C(O)=O
IUPAC Name oxalic acid dihydrate
Molecular Weight (g/mol) 126.06
Grade ACS
Density 1.65 g/cm3
Quantity 100 g
Physical Form Crystalline
Vapor Pressure <0.001 mm Hg
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.