Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalic Acid Dihydrate, MP Biomedicals™

Catalog No. ICN15255290 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15255290 500 g
02-152-55280 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15255290 Supplier MP Biomedicals, Inc Supplier No. 0215255290
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6153-56-6
Molecular Formula C2H6O6
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
PubChem CID 61373

Specifications

Color White
Quantity 500 g
Physical Form Crystalline
SMILES C(=O)(C(=O)O)O.O.O
IUPAC Name oxalic acid;dihydrate
Molecular Weight (g/mol) 126.064
Formula Weight 126.07
Chemical Name or Material Oxalic acid 2-hydrate, For ACS analysis
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.