Learn More
Oxalic Acid Dihydrate, Reagents
Specifications
Specifications
| Chemical Name or Material | Oxalic Acid Dihydrate |
| CAS | 6153-56-6 |
| Melting Point | 101°C to 102°C |
| Color | Colorless/White |
| Molecular Formula | C2H6O6, 2H2O |
| Synonym | Ethanedioic Acid dihydrate |
| Molecular Weight (g/mol) | 162.1 |
| Grade | Pure |
| Quantity | 3 kg |
| Packaging | Poly Bottle |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.