Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalic Acid dihydrate Suprapur™, 99.5%, MilliporeSigma™

Catalog No. M1004890100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1004890100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1004890100 Supplier MilliporeSigma Supplier No. 1.00489.0100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6153-56-6
Molecular Formula C2H6O6
Molecular Weight (g/mol) 126.06
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym Ethanedioic acid
IUPAC Name oxalic acid dihydrate
SMILES O.O.OC(=O)C(O)=O

Specifications

pH 1.5 (10 g/L, H2O)
Melting Point 98 to 100°C
Boiling Point 149 to 160°C (1013 hPa) (decomposition)
Density 1.65 g/cm3 (at 20°C)
Quantity 100 g
Flash Point 157°C (Decomposition)
Packaging Plastic Bottle
Assay Percent Range ≥ 99.5%
Vapor Pressure 0.000312 hPa (25°C)
Solubility Information > 100 g/L
Recommended Storage Store at 15 to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.