Learn More
FUJIFILM BIOSCIENCES INC Oxalic Acid Dihydrate
Supplier: FUJIFILM BIOSCIENCES INC 15600435
Oxalic Acid Dihydrate appearing as white crystals or crystalline powder soluble in water and ethanol with minimal solubility in diethyl ether Used as a general-purpose reagent reducing agent and bleaching agent as well as for the purification of compounds For research use only RUO