Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalic Acid TS, 6.3% (w/v), Ricca Chemical
SDP

Catalog No. 546632 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5466-32 1 L Natural Poly Bottle
5466-16 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5466-32 Supplier Ricca Chemical Company Supplier No. 546632
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 6153-56-6
Molecular Formula C2H6O6
Molecular Weight (g/mol) Mixture
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
PubChem CID 61373
IUPAC Name oxalic acid;dihydrate
SMILES C(=O)(C(=O)O)O.O.O

Specifications

pH 1.3 (0.1M)
Melting Point ∼0°C
Boiling Point ∼100°C
CAS Min % 92.0, 5.81
Color Colorless
CAS Max % 96.0, 6.05
Concentration 6.3% (w/v)
Quantity 1 L
Packaging Natural Poly Bottle
Physical Form Liquid
Specific Gravity ∽1.02 to 1.05
Solubility Information Infinite
Percent Purity 6.3% w/v
Grade Laboratory
Concentration or Composition (by Analyte or Components) 6.3% (w/v)
Chemical Name or Material Oxalic Acid, Test Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.