Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Oxaliplatin, Platinum Anticancer Agent, Solid, 50 mg

Catalog No. 502706944
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6944 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6944 Supplier Abcam Supplier No. AB14105450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diaminocyclohexane platinum anticancer agent that induces DNA cross-linking and blocks DNA synthesis.

  • Molecular weight: 397.29 g/mol
  • Purity: >99%
  • Form: Solid
  • Description: Diaminocyclohexane platinum anticancer agent that induces DNA cross-linking, blocks DNA synthesis, and induces apoptosis and cell cycle arrest.
  • Storage buffer: Constituents: 100% Oxaliplatin
  • Storage temperature: Store at +4°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule DNA
Shipping Condition Ambient
Content And Storage Store at 4°C.
Form Solid
Formulation 100% Oxaliplatin
Quantity 50 mg
For Use With (Application) Cancer research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.