Learn More
Enzo Life Sciences Oxatomide (10mg). CAS: 60607-34-3
SDP
Supplier:  Enzo Life Sciences BMLEI1140010
Encompass_Preferred
Suppresses PAF-induced bronchoconstriction in the guinea pig (ED50=0.9 mg/kg orally). Purity: ≥99% (HPLC). Solubility: Soluble in ethanol or DMSO. Long Term Storage: Ambient.
Catalog No. 50-201-0558
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.