Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxcarbazepine 98.0+%, TCI America™
SDP

Catalog No. O036325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O036325G 25 g
O03635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O036325G Supplier TCI America Supplier No. O036325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28721-07-5
Molecular Formula C15H12N2O2
Molecular Weight (g/mol) 252.273
MDL Number MFCD00865307
InChI Key CTRLABGOLIVAIY-UHFFFAOYSA-N
Synonym oxcarbazepine, trileptal, oxcarbamazepine, oxacarbazepine, oxcarbazepina, oxcarbazepinum, timox, oxcarbazepinum inn-latin, oxcarbazepina inn-spanish, oxcarbazepine inn
PubChem CID 34312
ChEBI CHEBI:7824
IUPAC Name 5-oxo-6H-benzo[b][1]benzazepine-11-carboxamide
SMILES C1C2=CC=CC=C2N(C3=CC=CC=C3C1=O)C(=O)N

Specifications

Melting Point 216°C
Quantity 25 g
Formula Weight 252.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Oxcarbazepine
RTECSNumber HN8445000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.