Learn More
JDM SCIENTIFIC LLC OXFORD BENCHMATE MULTI-POSITIO
Supplier:  JDM SCIENTIFIC LLC MHS10
Encompass_Preferred
Oxford BenchMate Multi-Position Magnetic Hotplate Stirrer - 10 Position Magnetic Hotplate Stirrer, Speed range - 300-1500 rpm, Max temp 120C, with 1 Stir Bar. 110/230 V, 50/60Hz.
Catalog No. NC2127787
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.