Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Oxidation-Reduction Potential Sensor
SDPSCI_ED

Catalog No. S16044ND Shop All Vernier Software Products
Change view
Click to view available options
Temperature Range (Metric):
0°C to 60°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric)
S16044ND 0°C to 60°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16044ND Supplier Vernier™ Supplier No. ORPBTA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

ORP Electrode BNC, Electrode Amplifier, pH storage solution

Requires

Requires an interface and software, sold separately

The Oxidation-Reduction Potential (ORP) Sensor measures the ability of a solution to act as an oxidizing or reducing agent. Use the ORP Sensor to measure the oxidizing ability of chlorine in swimming pools or to determine when the equivalence point has been reached in an oxidation-reduction reaction.

Type: Sealed, gel-filled, epoxy body, Ag/AgCl reference Storage solution: pH-4/KCl solution (10 g KCl in 100 mL buffer pH-4 solution) Cable: 1 meter coaxial cable with BNC connector Redox potential range: -450 to +1100 mV ORP element: 99% pure platinum band sealed on a glass stem Temperature range: 0-60ºC 12 mm OD Impedance: ∼20 kΩ at 25ºC

Warranty and Services

5-year limited warranty

Specifications

Type Sealed, Gel-filled, Epoxy Body, Ag/AgCl Reference
Includes ORP Electrode BNC, Electrode Amplifier, pH storage solution, user manual
Interface Required
For Use With (Application) Educational use only
Range Redox potential range -450 to 1100 mV
Temperature Range (Metric) 0°C to 60°C
Diameter (Metric) Outside 12 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.