Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversey™ Oxivir™ Tb Disinfectant Solution

Catalog No. 19855004 Shop All Oakton Products
Change view
Click to view available options
Volume (English):
1 gal.
55 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-855-004 55 gal.
19-855-003 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-855-004 Supplier Diversey™ Supplier No. 7890086
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tough on pathogens, not on people

  • Disinfects surfaces in less than one minute
  • Ready-to-use, one-step disinfectant saves time
  • Gentle formula suitable for daily use
  • Non-irritating to skin and eyes
  • Penetrates through the toughest soils
  • Kills SARS-CoV-2 (COVID-19 Virus)

Specifications

Product Type Disinfectant Solution
Color-coded No
Form Liquid
pH 3
Sterility Non-sterile
Composition Benzyl Alcohol, Hydrogen Peroxide, Dodecylbenzene Sulfonic Acid
For Use With (Application) General Virucide, Bactericide, Tuberculocide, Fungicide, Sanitizer
Quantity 1 Drum
Size 55 gal.
Volume (English) 55 gal.
Volume (Metric) 208 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.