Learn More
Description
Oxoid Agar Technical (Agar No. 3) is a general-purpose bacteriological-grade agar.
- Normal working concentration 1.2%
Specifications
Specifications
| Description | Agar Tech (agar No 3) 5 Kg |
| Form | Powder |
| Product Type | Agar |
| Quantity | 5 kg |
For Laboratory Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.