Learn More
Description
Carry out microbiological assay of antibiotics with the seed layer of Thermo Scientific™ Oxoid™ Antibiotic Medium No.1 (Dehydrated). This is one of the most important media in antibiotic assay work and it is in a specially modified form to take advantage of the properties of Oxoid Agar No.1. (LP0011).
Certain agars have inhibitory properties towards some antibiotics, particularly streptomycin, kanamycin, polymyxin B and neomycin1. Oxoid Antibiotic Medium No.1 does not share these inhibitory properties and is therefore especially suited for antibiotic assay work.
Antibiotic Medium No.1 is a recommended medium for the seed layer in the preparation of plates for the microbiological assay of antibiotics.
- The medium is used as a seed agar with Micrococcus flavus for the plate assay of bacitracin; with Sarcina lutea for the plate assay of chloramphenicol and with Staphylococcus aureus for the assay of kanamycin sulphate, penicillin G, sodium methicillin and sodium oxacillin.
- Employed as a base agar in the assay of the number of drugs like chloramphenicol, kanamycin sulphate, colistin sulphate, sodium methicillin, sodium oxacillin and vancomycin hydrochloride.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: AOAC, FDA, USDA, ISO. |
| Description | Antibiotic Medium No. 1 |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Quantity | 500 g |
| Yield | For 18.5L medium |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.