Learn More
Thermo Scientific™ Antibiotic Medium No. 3 Assay Broth (Dehydrated)
Shop All Thermo Scientific ProductsDescription
Carry out serial dilution assay of penicillin and other antibiotics with Thermo Scientific™ Oxoid™ Antibiotic Medium No. 3 Assay Broth (Dehydrated). Antibiotic Medium No. 3 Assay Broth is used as the broth medium in turbidimetric or serial dilution assay of a wide variety of antibiotics with Staphylococcus aureus.
Antibiotic Medium No. 3 Assay Broth is used as the broth medium in turbidimetric or serial dilution assay of a wide variety of antibiotics with Staphylococcus aureus.
Taking advantage of modern technology, this medium is based on the original `Penassay Broth.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
| Description | Antibiotic Medium No. 3 |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Quantity | 500 g |
| Yield | For 28.6L medium |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.