Learn More
Description
Selectively enrich Arcobacter species with this medium when used with CAT or CCDA selective supplement.
Selectively enrich Arcobacter species with Thermo Scientific™ Oxoid™ Acrobacter Broth Base (Dehydrated) when used with either CAT Selective Supplement(SR0174) or CCDA Selective Supplement (SR0155).
Arcobacter Broth is intended for use with Cefoperazone, Amphotericin B, Teicoplanin (CAT) Selective Supplement (SR0174) as a selective enrichment broth for the growth of Arcobacter species and with the more selective CCDA (SR0155) for the selective enrichment of Arcobacter butzleri.
Arcobacters are micro aerophilic, Gram-negative rods, which were formerly classified as Campylobacter1. Arcobacter butzleri, Arcobacter cryaerophilus, and Arcobacter skirrowii have been associated with disease in humans2,3, and typically are isolated from fecal samples.
- Specifically designed to provide the ideal growth conditions for Arcobacter
- Selective for Arcobacter species.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
| Description | Arcobacter Broth Base |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Quantity | 500 g |
| Yield | For 20.8L medium |
For Laboratory Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.