Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Brilliance™ Bacillus Cereus Agar Base (Dehydrated)

Catalog No. R661362 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R661362 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R661362 Supplier Thermo Scientific™ Supplier No. CM1036B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isolate and differentiate Bacillus cereus from food samples with this chromogenic and selective medium.

Isolate and differentiate Bacillus cereus from food samples with chromogenic Thermo Scientific™ Brilliance™ Bacillus Cereus Agar Base (Dehydrated).

BrillianceBacillus cereus Agar incorporates the chromogenic substrate 5-bromo-4-chloro-3-indolyl-ß-glucopyranoside, which is cleaved by the enzyme ß-glucosidase present in Bacillus cereus resulting in the formation of blue/green colonies. Combination of Polymixin B and Trimethoprim inhibit most Gram-positive bacteria and Gram-negative bacteria other than Bacillus cereus respectively.

Bacillus cereus, a Gram-positive, aerobic, spore-forming rod-shaped bacterium, is widely distributed in nature. It is readily isolated from soil, dust, cereal crops, vegetation, animal hair, fresh water and sediments. Therefore, it is not surprising to find the organism associated with virtually every raw agricultural commodity.

The role of Bacillus cereus in food poisoning, particularly from the consumption of contaminated rice, is now well documented1,2,3. The organism has also been implicated in eye infections4,5 and a wide range of other conditions, including abscess formation, meningitis, septicemia, and wound infection.

  • Easy-to-read: cleavage of the chromogenic substrate the enzyme ß-glucosidase present in Bacillus cereus results in the formation of blue/green colonies.
  • Selective: a combination of Polymixin B and Trimethoprim make the medium selective for Bacillus cereus.

Brilliance Bacillus cereus Agar incorporates the chromogenic substrate 5-bromo-4-chloro-3-indolyl-ß-glucopyranoside, which is cleaved by the enzyme ß-glucosidase present in Bacillus cereus resulting in the formation of blue/green colonies. Polymixin B inhibits most Gram-negative organisms and some Gram-positive organisms including some Bacillus other than Bacillus cereus. Trimethoprim, which is also added to the medium, blocks folic acid synthesis necessary for DNA production and is active against many Gram-positive bacteria including Staphylococcus aureus, Enterococcus spp. and some non-cereus Bacillus species. The combination of these two antibiotics has been shown to be more effective than the use of polymyxin B alone3.

Because Bacillus thuringiensis is biochemically identical to Bacillus cereus, it will also grow as blue/green colonies on this medium. Bacillus thuringiensis is known primarily as an insect pathogen, but it has also been reported to have been linked to some human gastroenteritis outbreaks4.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Brilliance Bacillus Cereus Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 12.2L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.