Learn More
Thermo Scientific™ Brilliance™ E. coli/Coliform Selective Medium (Dehydrated)
Shop All Thermo Scientific ProductsDescription
Detect and enumerate Escherichia coli and other coliforms in food and water samples with this selective, chromogenic medium.
Detect and enumerate Escherichia coli and other coliforms in food and water samples with selective, chromogenic Thermo Scientific™ Brilliance™ E. coli / Coliform Selective Agar. The agar base uses two chromogens to differentiate between Escherichia coli and other coliforms. This results in purple E. coli, as they are able to cleave both chromogens while other coliforms, giving pink colonies.
The recovery and enumeration of Escherichia coli and coliforms are important indicators of environmental and food hygiene. Use Brilliance E. coli / Coliform Selective Agar for detection and enumeration of Escherichia coli and other coliforms in food and water samples.
- Easy-to-read: purple E. coli colonies are easy to differentiate from pink colonies of other coliforms
- Selective: due to the presence of selective agent sodium lauryl sulphate the medium is selective against Gram-positive organisms.
Brilliance E. coli / Coliform Selective Agar contains two chromogenic agents:
Rose-Gal: detects ß-galactosidase activity
X-Glu: detects ß-glucuronidase activity
Detection of ß-glucuronidase activity is widely used to differentiate Escherichia coli, as this is present in E. coli but not in other members of the coliform group. As most coliforms are lactose-positive, ß-galactosidase activity, encoded by the lacZ gene, is then used to differentiate this group from other organisms able to grow on the selective medium. This results in purple E. coli, as they are able to cleave both chromogens with other coliforms, giving pink colonies, as they cleave only the galactoside chromogen.
The medium also contains sodium lauryl sulphate which acts as a selective agent, inhibiting the growth of Gram-positive organisms. Due to the addition of tryptophan, the rapid confirmation of E. coli by the formation of indole using a spot indole test is possible.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
| Description | Brilliance E. coli/Coliform Selective Medium |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Quantity | 500 g |
| Yield | For 17.8L medium |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.