Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Brilliance™ Listeria Selective Supplement

Catalog No. SR0227E
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SR0227E 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SR0227E Supplier Thermo Scientific™ Supplier No. SR0227E

Brilliance Listeria Selective Supplement is added to Listeria Agar Base to isolate/differentiate Listeria spp. in food samples.

Brilliance Listeria Selective Supplement is added to Brilliance Listeria Agar Base, Mfr. No. CM1080E and Mfr. No. CM1080B, a chromogenic medium for the isolation and differentiation of Listeria spp. in food samples.

Listeria monocytogenes is the most common pathogenic Listeria spp. and has been shown to be pathogenic to both man and animals. Some Listeria ivanovii strains also possess lecithinase activity and, although Listeria ivanovii are primarily pathogenic to animals, there are strains which have been shown to cause infection in humans. Brilliance Listeria Agar uses the chromogen X-glucoside for presumptive identification of Listeria spp. This chromogen is cleaved by β-glucosidase which is common to all Listeria species. Other organisms that possess this enzyme, such as enterococci, are inhibited by the selective agents within the medium; lithium chloride, polymyxin B and nalidixic acid, while amphotericin inhibits the growth of any yeasts and moulds present in the sample. Listeria monocytogenes and pathogenic Listeria ivanovii are then further differentiated by their ability to produce the phospholipase enzyme, lecithinase. This enzyme hydrolyses the lecithin in the medium, producing an opaque white halo around the colony. Brilliance Listeria Agar is a modification of the formulation described by Ottaviani and Agosti. As in the original formulation, the medium is designed to identify Listeria spp. based on their utilization of a chromogenic substrate. However, in this modification, the pathogenic Listeria spp. are then further differentiated by the detection of lecithinase (phosphotidylcholine phospholipase C (PCPLC) activity, rather than phosphotidylinositol phospholipase C (PIPLC) activity. Both enzymes, PCPLC and PIPLC, are associated with virulence in Listeria spp., and, therefore, the presence of either enzyme is a useful indicator of pathogenicity.
  • Add to Brilliance Listeria Selective Agar Base, Part No. CM1080B
  • Each vial supplements 500mL of medium
ONE Broth-Listeria Base and ONE Broth Selective Supplement are for Laboratory Use Only.
  • ONE Broth-Listeria Base is designated as “Irritant” and ONE Broth-Listeria Selective Supplement is designated as “Harmful.” For a full Material Safety Data Sheet (MSDS), contact your customer service or sales representative.
  • Do not use ONE Broth-Listeria Base or ONE Broth-Listeria Selective Supplement beyond the stated expiration dates, or if the products show any sign of deterioration.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Description Brilliance Listeria Selective Supplement
    Form Freeze-dried
    Product Type Selective Supplement
    Quantity 10/Pk.
    Yield Each vial for 500mL medium
    Species Listeria
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.