Learn More
Thermo Scientific™ Oxoid™ Buffered Listeria Enrichment Broth
Description
Oxoid Buffered Listeria Enrichment is selective enrichment medium for the detection of Listeria monocytogenes.
- Intended for use with samples of fermented products and as an alternative method for enrichment of environmental specimens
- Also available Listeria Selective Supplement, Part No. SR0140E or SR0143E; Modified Listeria Selective Enrichment Supplement, Part No. SR0206E or SR0206B; and Listeria Selective Enrichment Supplement, Part No. SR0141E
Specifications
Specifications
| Content And Storage | Buffered Listeria Enrichment Broth should be stored tightly capped in the original container in a cool, dry place away from bright light. When stored as directed the medium will remain stable until the expiry date printed on the label. |
| Description | Buffered Listeria Enrichment Broth |
| Quantity | 500 g |