Learn More
Thermo Scientific™ Oxoid™ Buffered Listeria Enrichment Broth
Description
Oxoid Buffered Listeria Enrichment is selective enrichment medium for the detection of Listeria monocytogenes.
- Intended for use with samples of fermented products and as an alternative method for enrichment of environmental specimens
- Also available Listeria Selective Supplement, Part No. SR0140E or SR0143E; Modified Listeria Selective Enrichment Supplement, Part No. SR0206E or SR0206B; and Listeria Selective Enrichment Supplement, Part No. SR0141E
Specifications
Specifications
| Content And Storage | Buffered Listeria Enrichment Broth should be stored tightly capped in the original container in a cool, dry place away from bright light. When stored as directed the medium will remain stable until the expiry date printed on the label. |
| Description | Buffered Listeria Enrichment Broth; 2.5kg |
| Form | Powder |
| Product Type | Microbiology Media |
| Quantity | 2.5 kg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.