Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Buffered Peptone Water (Dehydrated)

Catalog No. OXCM0509T
Change view
Click to view available options
Quantity:
500 g
2.5 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0509T 5 kg
OXCM0509B 500 g
OXCM0509R 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. OXCM0509T Supplier Thermo Scientific™ Supplier No. CM0509T
Add to Cart
Add to Cart
This item is not returnable. View return policy

Pre-enrich food samples prior to the isolation of Salmonella species while allowing the resuscitation of sub-lethally injured cells with this medium.

Pre-enrich food samples prior to the isolation of Salmonella species using Thermo Scientific™ Oxoid™ Buffered Peptone Water (Dehydrated). Buffered Peptone Water is a non-selective pre-enrichment medium. It also provides conditions for resuscitation of cells that have been injured by processes of food preservation.

Low numbers and/or sub-lethally injured Salmonella spp. cells may be present in a test food sample. Pre-enrichment in non-selective Buffered Peptone Water allows the cells time to repair and multiply before being introduced into a selective culture, improving the chances of recovering Salmonella species from the sample.

  • Supports high growth and recovery rate: rich in nutrients, the medium produces high resuscitation rates and supports intense growth.
  • Helps recovery of sub-lethally damaged salmonellae: the phosphate buffer system of the medium prevents bacterial damage due to changes in pH of the medium and helps the recovery of sub-lethally damaged salmonellae that may be sensitive to low pH.

It was noted by Edel and Kampelmacher1 that sub-lethal injury to salmonellae may occur in many food processes. In a survey involving isolation of salmonellae from meat that had been artificially contaminated with sub-lethally injured organisms, pre-enrichment in buffered peptone water at 37°C for 18 hours before selection in Brilliant Green Tetrathionate Bile Broth showed superior results compared with a direct selection method.

Sadovski2 reported that, in experiments involving isolation of salmonellae from frozen vegetables, the rapid drop in pH when using lactose broth3 as a pre-enrichment medium was detrimental to the recovery of salmonellae. This was due to the enhanced sensitivity to low pH of freeze-injured salmonellae which may contaminate frozen vegetables. Pre-enrichment with Buffered Peptone Water maintained a high pH over a period of 24 hours of incubation. The vegetable tissue has a low buffering capacity and the medium overcame this problem.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description Buffered Peptone Water
Form Powder
Product Type Pre-enrichment Medium
Quantity 5 kg
Yield For 250.0L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.