Learn More
Thermo Scientific™ Buffered Sodium Chloride Peptone Solution EP/USP/JP/BP (Dehydrated)
Shop All Thermo Scientific ProductsDescription
Carry out microbial limit testing by harmonized methodology of pharmaceutical products in accordance with USP/EP/BP/JP with this medium.
Carry out microbial limit testing by harmonized methodology of pharmaceutical products in accordance with USP/EP/BP/JP1,2,3 with Thermo Scientific™ Oxoid™ Buffered Sodium Chloride Peptone Solution (Dehydrated). The combination of phosphate buffer, sodium chloride and peptone increases the viability of sensitive microorganisms by providing an isotonic environment. This is of particular importance when trying to recover cells that may be stressed or sensitive to osmotic pressure.
Use Buffered Sodium Chloride Peptone Broth for the dilution of samples in the analysis of non-sterile products for microbial contaminants.
- Conforms to the harmonized Microbial Limit Testing chapters from the EP/USP/BP/JP1,2,3
- Maintains cell viability during the dilution process.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: EP. |
| Description | Buffered Sodium Chloride Peptone Solution |
| Form | Powder |
| Product Type | Dehydrated Peptone |
| Quantity | 500 g |
| Yield | For 34.2L medium |
For Laboratory Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.