Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ Campylobacter Agar Base

Catalog No. OXCM0689B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0689B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0689B Supplier Thermo Scientific™ Supplier No. CM0689B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Oxoid Campylobacter Agar Base isolates Campylobacter jejuni and Campylobacter coli.

Oxoid Campylobacter Agar Base is used for the isolation of Campylobacter jejuni and Campylobacter coli from human, animal, avian and environmental specimens.

Also available Campylobacter Selective Supplement (Blaser-Wang), Part No. SR0098E NMKL; Bolton Broth Selective Supplement, Part No. SR0183E; Modified Bolton Broth Selective Supplement, Part No. SR0208E; Modified Preston Campylobacter Selective Supplement, Part No. SR0204E; and Campylobacter Selective Supplement Preston, Part No. SR0117E.
TRUSTED_SUSTAINABILITY

Specifications

Description Campylobacter Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 13.5L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.