Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Cetrimide Agar (USP, EP) (Dehydrated)

Catalog No. OXCM0579B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0579B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0579B Supplier Thermo Scientific™ Supplier No. CM0579B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selectively isolate and differentiate Pseudomonas aeruginosa from a range of samples with this medium while conforming to International Pharmacopoeia.

Selectively isolate and differentiate Pseudomonas aeruginosa from a range of samples using Thermo Scientific™ Oxoid™ Cetrimide Agar (USP, EP) (Dehydrated). The medium contains magnesium chloride and potassium sulphate for improved pigment production and recovery of Pseudomonas aeruginosa while strongly suppressing Klebsiella, Proteus and Providencia spp. The medium conforms to European Pharmacopoeia for Microbial Limit Tests1.

Cetrimide Agar (USP, EP) is a modification of the original medium described by Brown and Lowbury2 for the selective isolation and differentiation of Pseudomonas aeruginosa from a range of samples. Cetrimide is a quaternary ammonium compound with bactericidal activity against a broad range of Gram-positive organisms and some Gram-negative organisms.

  • Pseudomonas aeruginosa forms fluorescent green colonies while the growth of Klebsiella, Proteus and Providencia is strongly suppressed.
  • Enhanced pigment production aids identification
  • Conforms to U.S. Pharmacopeia3 and European Pharmacopoeia1 standards for Microbial Limit Tests.
  • The formulation is in line with the A.O.A.C. guidelines4 for isolation of Pseudomonas aeruginosa from cosmetics and in A.O.A.C. method5 for testing disinfectants on hard surfaces.

Pseudomonas aeruginosa produces a number of water soluble iron chelators, including the yellow-green or yellow-brown fluorescent pyoverdin. When pyoverdin combines with the blue water-soluble pyocyanin, the bright green colour characteristic of Pseudomonas aeruginosa is created. The addition of magnesium chloride and potassium sulphate enhances the production of these chelators.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AOAC, EP, USP.
Description Pseudomonas Cetrimide Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 11.0L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.