Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Chromogenic Cronobacter Isolation Agar (Dehydrated)

Catalog No. CM1122B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CM1122B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CM1122B Supplier Thermo Scientific™ Supplier No. CM1122B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Differentiate and enumerate Cronobacter sakazakii from infant formula and other food samples with this chromogenic medium.

Differentiate and enumerate Cronobacter sakazakii from infant formula and other food samples with Thermo Scientific™ Oxoid™ Chromogenic Cronobacter Isolation Agar (Dehydrated). Chromogenic Cronobacter Isolation Agar (CCI) is an enhanced formulation of Brilliance™ Cronobacter Sakazakii Agar (DFI) with improved specificity and colony coloration for the detection of Cronobacter.

Cronobacter sakazakii is a Gram-negative rod-shaped bacterium that rarely causes disease in healthy adults. It has been implicated in outbreaks of disease in premature infants (neonates). Research suggests that neonates, or those infants who have other medical conditions, are more susceptible to this infection. Most reported cases of infection are severe, including sepsis (bacteria in the blood, meningitis, or necrotizing enterocolitis (severe intestinal infection).

Use Chromogenic Cronobacter Isolation Agar for differentiation and enumeration of Cronobacter sakazakii from infant formula and other food samples.

  • Selective: due to the presence of desoxycholate the medium is selective against most Gram-positive organisms.
  • Easy-to-read: Cronobacter sakazakii produces blue-green colonies on this medium.

Cronobacter spp. have been isolated at low levels from powdered infant formulae, and the organisms’ high tolerance to desiccation provides a competitive advantage in the dry environments of milk powder factories, increasing the risk of product contamination1.

Chromogenic Cronobacter Isolation (CCI) agar is an optimisation of the original Brilliance™ Enterobacter Sakazakii Agar (DFI), developed by Iversen et al. in 20043. The concentration of the X-α-glucoside chromogen was increased to improve the blue-green colouration of Cronobacter strains with weak glucosidase activity. Sodium thiosulphate and ammonium iron (III) citrate are included to differentiate hydrogen sulphide-producing Enterobacteriaceae, which are coloured grey/brown, and sodium deoxycholate is added to inhibit the growth of Gram-positive competitors.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Chromogenic Cronobacter Isolation Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 16.3L medium
Species Cronobacter

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.