Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Clostridium Difficile Agar Base (Dehydrated)

Catalog No. OXCM0601B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0601B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0601B Supplier Thermo Scientific™ Supplier No. CM0601B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate Clostridium difficile from fecal specimens with this medium when used with Culture Media Supplement (SR0096).

Isolate Clostridium difficile from fecal specimens with Thermo Scientific™ Oxoid™ Clostridium Difficile Agar Base (Dehydrated) when used with Culture Media Supplement (SR0096). The combination of Clostridium Difficile Agar Base and the Culture Media Supplement (SR0096) is based on the formulation proposed by George et al.1 The selective agents D-cycloserine (500μg/ml) and cefoxitin (16μg/ml) inhibit growth of the majority of non target organisms which may be found in large numbers in fecal samples.

Clostridium difficile was first isolated in 1935 by Hall and O’Toole2 who proposed the name `difficile’ because it was very difficult to isolate. Clostridium difficile is a toxin-producing, spore forming anaerobic gram-positive rod, which causes gastrointestinal infections ranging in severity from asymptomatic colonization to severe diarrhea, pseudomembranous colitus (PMC), toxic megacolon and colonic perforaton.

Clostridium Difficile Agar Base is used for the primary isolation of C. difficile from fecal specimens. The medium composition is designed so as to obtain luxuriant growth of C. difficile.

  • Selective: the selective agents D-cycloserine (500μg/mL) and cefoxitin (16μg/mL) inhibit growth of the majority of Enterobacteriaceae, as well as Streptococcus fecalis, staphylococci, Gram-negative non-sporing anaerobic bacilli and Clostridia (except Clostridium difficile) which may be found in large numbers in fecal samples.
  • Easy-to-read: C. difficile forms grey-white colored colonies.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Clostridium difficile Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 7.2L medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.