Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Cold Filterable Vegetable Peptone Broth (Dehydrated)

Catalog No. VG0104B
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
VG0104B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. VG0104B Supplier Thermo Scientific™ Supplier No. VG0104B
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Carry out microbiological Media Fill Trials (MFT) in the pharmaceutical industry with this gamma-irradiated medium.

Carry out microbiological Media Fill Trials (MFT) in the pharmaceutical industry with gamma-irradiated Thermo Scientific™ Oxoid™ Cold Filterable Vegetable Peptone Broth (Dehydrated). The highly nutritious general purpose medium conforms to the formulation stated for MFT in the European1, British2, US3 and Japanese4 Pharmacopoeias.

Cold Filterable Vegetable Peptone Broth can be substituted for the powdered components that go into making aqueous drug products to provide an index of the level of microbial contamination occurring in aseptic manufacture.

‘Sterile for use’ liquid drugs often contain heat-sensitive components which means that terminal sterilization by autoclaving is not an option. Sterilization by filtering (for soluble liquids) followed by filling under aseptic conditions is the method for preparation of these types of drug. The purpose of the Media Fill Trials is to provide a measure of the likelihood of microbiological contamination arising in a particular aseptic process.

Packs of Cold Filterable Vegetable Peptone Broth have been given a sterilizing dose of gamma-irradiation (minimum 30KGy) validated as a lethal dose for all yeasts, molds, bacterial spores and mycoplasma.

  • Gamma-irradiated to ensure that the medium is free from all yeasts, molds and bacteria, including bacterial spores and mycoplasmas, prior to use.
  • Screened and selected to ensure a highly filterable solution that has a known Vcap value. This allows the maximum filterable volume to be calculated before starting a MFT.

Aqueous liquid products that are required to be sterile but cannot be terminally sterilized due to the heat-sensitive nature of one or more component, may be produced by filtering-sterilizing the dissolved solution and maintaining sterility by filling and closing the product under aseptic conditions.

The purpose of MFT is to provide a measure of the likelihood of microbiological contamination occurring in a particular aseptic process. Cold Filterable Vegetable Peptone Broth can be used as a substitute for filter-sterilized drug products and is processed in a manner identical to that in which the product would be processed i.e. filtering, filling and closing. The medium is then incubated and the number of contaminated units is scored versus those that are un-contaminated. A decision following pre-determined guidelines can be made based on the proportion of contaminated units and the identity of the microorganisms recovered5

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description Cold Filterable Vegetable Peptone Broth
Form Powder
Media Type Vegetable Peptone Broth
Product Type Nutrient Broth
Quantity 500 g
Yield For 16.1L medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.