Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ Corn Meal Mycological Agar

Catalog No. OXCM0103B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0103B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0103B Supplier Thermo Scientific™ Supplier No. CM0103B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Maintain the fungal stock culture and observe chlamydispore production by Candida albicans with this mycological medium

Maintain the fungal stock culture and observe chlamydospore production by Candida albicans with Thermo Scientific™ Oxoid™ Corn Meal Mycological Agar (Dehydrated). Microscopic examination of Candida albicans on this medium shows the characteristic chlamydospore production which is an accepted criterion for the identification of this species. Corn Meal Mycological Agar, being a nutritionally impoverished medium, can be employed for the maintenance of stock cultures of fungi, especially the black pigmented varieties.

Use Corn Meal Mycological Agar for chlamydospore production by Candida albicans for the maintenance of fungal stock cultures.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Corn Meal Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 29.4L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.