Learn More
Thermo Scientific™ Czapek Dox Liquid Medium (Modified) (Dehydrated)
Shop All Thermo Scientific ProductsDescription
Cultivate those fungi and bacteria which can utilize sodium nitrate as the sole source of nitrogen using Thermo Scientific™ Oxoid™ Czapek Dox Liquid Medium (Modified) (Dehydrated).
Use Czapek Dox Liquid Medium (Modified) for cultivation of those fungi and bacteria which can utilize sodium nitrate as the sole source of nitrogen.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
| Description | Czapek Dox Liquid Medium |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Quantity | 500 g |
| Yield | For 15.0L medium |
For Laboratory Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.