Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Reinforced Clostridial Agar (RC Agar) (Dehydrated)

Catalog No. OXCM0151B Shop All Thermo Scientific Products
Change view
Click to view available options
Form:
Powder
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form Quantity
OXCM0151B Powder 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0151B Supplier Thermo Scientific™ Supplier No. CM0151B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enumerate and cultivate clostridia and other anaerobes occurring in food and pathological samples with this solid medium.

Cultivate and enumerate anaerobes, especially Clostridium species, with Thermo Scientific™ Oxoid™ Reinforced Clostridial Agar (RC Agar) (Dehydrated). A solid, highly nutritious medium, it can be used as an enrichment medium to optimize the growth of clostridia and other anaerobes, lactobacilli, and many other species of bacteria.

Use Reinforced Clostridial Agar for the isolation and cultivation of anaerobic organisms occurring in a variety of habitats, including food and pathological specimens.

Reinforced Clostridial Agar is a solid version of Thermo Scientific Oxoid Reinforced Clostridium Medium suitable for the cultivation and enumeration of clostridia and other anaerobes. Reinforced Clostridial Agar does not differ significantly in performance from the pork-starch-pea agar of Anderson1for the count of anaerobes.

It is employed for the estimation of clostridia in food—see below (also Barnes et al.2 and Angelotti3). Attenborough and Scarr4employed RC Agar, in conjunction with membrane filters, for the count of Clostridium thermosaccharolyticum in sugar.

Reinforced Clostridial Agar is also frequently employed for the investigation of intestinal flora: Perry et al.5for investigation of bovine rumen streptococci; Williams Smith & Crabb6 used the Oxoid medium with added sodium chloride or blood for counts on human or animal feces; Barnes & Goldberg7 employed the Oxoid medium with added chlortetracycline hydrochloride or sodium azide and ethyl violet, for the examination of poultry feces; the Oxoid medium was also used by Goldberg et al.8 for the examination of poultry fecal samples; Williams Smith9 employed Oxoid Reinforced Clostridial Agar with added blood for the ‘total’ and Lactobacillus count of human and animal feces; with added blood and neomycin for determination of Bacteroides, anaerobic Gram-negative cocci, ‘total’ streptococci, sporeformers, yeasts, and ‘actino’ types.

Sneath10 used Oxoid RC Agar and other media for the anaerobic count of microorganisms from soil samples up to approximately 300 years old. Gregory et al.11 also employed the Oxoid medium for the estimation of anaerobes in molding hay.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 10°C to 30°C
Description Reinforced Clostridial Agar (RC Agar) (Dehydrated)
Form Powder
Media Type Reinforced Clostridial Agar
Product Type Agar
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.