Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Reinforced Clostridial Medium (RCM) (Dehydrated)

Catalog No. OXCM0149B
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Enumerate and cultivate clostridia and other anaerobes occurring in food and pathological samples with this medium.

Cultivate and enumerate clostridia and other anaerobes occurring in food and pathological samples with Thermo Scientific™ Oxoid™ Reinforced Clostridial Medium. A semi-solid, highly nutritious medium can be used as an enrichment medium to optimize the growth of clostridia before sub-culturing to solid agar.

Use Reinforced Clostridial Medium (RCM) for the isolation and cultivation of anaerobic organisms occurring in a variety of habitats, including food and pathological specimens.

  • Superior fertility: the medium produced the highest viable count when compared to other media tested by Hirsch & Grinstead 1
  • Differential: can be made differential for sulphite-reducing clostridia by the addition of sodium sulphite and ferric citrate2

Reinforced Clostridial Medium (RCM) was designed by Hirsch & Grinstead1 for the cultivation and enumeration of clostridia. They showed that the medium was more fertile and enabled growth to be initiated from small inocula more readily than five other media tested. In a further comparison, the highest viable count obtainable was the criterion used, and again, RCM proved superior. Compared with the spleen infusion medium of Mundt & Jones3, RCM gave somewhat higher counts (Gibbs & Hirsch4).

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 10°C to 30°C
Description Reinforced Clostridial Medium (RCM) (Dehydrated)
Form Powder
Media Type Reinforced Clostridial Medium
Product Type Dehydrated Culture Media
Quantity 500 g

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.