Learn More
Description
Isolate, enumerate and differentiate coliforms in dairy products with differential Thermo Scientific™ Oxoid™ Desoxycholate Agar. The medium is also used for the isolation of enteric pathogens from rectal swabs, feces or other specimens. Differentiation of coliforms is based on the fermentation of lactose. Lactose fermenting bacteria form pink colored colonies while non-lactose fermenters form straw colored colonies.
Desoxycholate Agar is a differential medium for the direct count of coliforms in dairy products (American Public Health Association1).
The medium is used in a `pour-plate’ technique or as a surface inoculated medium. A thin layer of uninoculated desoxycholate agar poured over the surface of a gelled `pour-plate’ assists subsequent counting.
Technique: Enumeration of Coliforms in Milk and Cream (APHA1) 1. Pipette 1-4ml of the sample (or decimal dilution of the sample) into a sterile Petri dish. 2. Cool freshly prepared Desoxycholate Agar to 42-44°C and add 10-20ml to each dish. 3. Mix the contents of the dishes by gentle tilting and rotation. 4. Allow the plates to solidify and pour on an overlay of 3-4ml of uninoculated Desoxycholate Agar. 5. When the overlay has set, invert the plates and incubate them for 18-24 hours at 35°C. 6. Count all dark red colonies measuring at least 0.5mm in diameter and calculate the number of coliform colonies per millilitre or gram of original sample. Isolation of Enterobacteriaceae It is advisable to use Desoxycholate Agar in parallel with other plating media for this purpose. Lightly inoculate a Desoxycholate Agar plate with faeces, rectal swab, or enrichment culture. Incubate for 18-24 hours at 35°C and examine. Non-lactose fermenters of enteric origin form colourless colonies. Non-lactose fermenters which are not of enteric origin are generally inhibited by the sodium desoxycholate in the medium. Identify suspect colonies in the usual manner. The selective agents in this medium may have inhibitory effects on the colony forming ability of Escherichia coli: the degree of inhibition varies with strains. The inhibition may be more pronounced when the cells are enumerated by pour plate rather than surface methods2.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Description | Desoxycholate Agar |
| Form | Powder |
| Product Type | Agar |
| Quantity | 500 g |
| Yield | For 11.1L medium |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.