Learn More
Description
Isolate and recover intestinal pathogens using this medium.
Isolate and recover intestinal pathogens using Thermo Scientific™ Oxoid™ Desoxycholate Citrate Agar (Dehydrated).
The medium is pale pink and clear. Lactose fermenting organisms produce pink colonies and may be surrounded by a zone of precipitated desoxycholic acid. The colonies of non-lactose fermenters are colorless, and due to their alkaline reaction, they are surrounded by a clear orange-yellow zone of medium.
Desoxycholate Citrate Agar is a modification of Leifson medium1, for the isolation and maximum recovery of intestinal pathogens. It is less selective and inhibiting than Desoxycholate Citrate Agar (Hynes) CM0227.
- Easy-to-read: provides an opaque background to easily discern the clearing produced by alkali-producing pathogens.
- Preferable when shigellae are being sought as well as salmonellae2.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Description | Desoxycholate Citrate Agar |
| Form | Powder |
| Product Type | Agar |
| Quantity | 500 g |
| Yield | For 10.3L medium |
For Laboratory Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.