Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Dextrose Tryptone Agar (Dehydrated)

Catalog No. OXCM0075B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0075B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0075B Supplier Thermo Scientific™ Supplier No. CM0075B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detect and enumerate flat-sour thermophiles and mesophiles in food products with this medium.

Detect and enumerate flat-sour thermophiles and mesophiles in food products with Thermo Scientific™ Oxoid™ Dextrose Tryptone Agar (Dehydrated). The medium, evolved as a result of several years of research by Williams1 is most suitable for the cultivation and enumeration of the thermophilic bacteria causing flat-sour spoilage of canned food.

Canned foods are more often prone to flat sour spoilage due to contamination by either mesophilic or thermophilic aerobic spore formers.

Dextrose Tryptone Agar is a bacteriologically controlled medium for the detection and enumeration of thermophilic and mesophilic organisms in food products. Its use for routine cultural purposes is recommended by Cameron2 and the Association of Official Analytical Chemist3. Dextrose Tryptone Agar is also recommended by:

  1. Tanner4 for the examination of canned food, sugar, and starch for thermophilic bacteria of the Bacillusstearo thermophilus type (i.e. flat-sour spoilage bacteria).
  2. The American Public Health Association5 for the enumeration of mesophilic and thermophilic aerobic bacteria in sweetening agents used in frozen dairy foods.
  3. The National Canners Association6 for determination of the total plate and flat-sour count of thermophilic bacteria spores in ingredients, such as sugar and starch.
  4. The American Public Health Association7 for the enumeration of mesophilic organisms and flat-sour spores in sugars, starches and other complex carbohydrates; and for the enumeration of flat-sour thermophiles in cereals and cereal products, dehydrated fruits and vegetables, and spices.
  5. Baumgartner and Hersom8 for the examination of low and medium-acid canned food (above pH 4.5) for flat-sour thermophiles, mesophilic aerobes, and facultative anaerobes

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description Dextrose Tryptone Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 18.5L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.