Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Dextrose Tryptone Broth (Dehydrated)

Catalog No. OXCM0073B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0073B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0073B Supplier Thermo Scientific™ Supplier No. CM0073B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cultivate and detect organisms causing spoilage in canned foods and other products with this medium.

Cultivate and detect organisms causing spoilage in canned foods and other products with Thermo Scientific™ Oxoid™ Dextrose Tryptone Broth (Dehydrated). Organisms which produce acid from dextrose, such as Bacillus stearothermophilus and other 'flat-sour' organisms, are detected by the color change of the medium from purple to yellow.

Canned foods are more often prone to flat sour spoilage due to contamination by either mesophilic or thermophilic aerobic spore formers.

Dextrose Tryptone Broth is recommended by the American Public Health Association1 and Baumgartner and Hersom2 for the bacterial examination of low and medium acid canned foods (pH 4.5 and above). Both methods include inoculation of 10mL amounts of the broth with one or two grams of the food product. For food products in this pH range, the suggested procedure is aerobic cultivation in Dextrose Tryptone Broth in parallel with anaerobic cultivation in other media.

TRUSTED_SUSTAINABILITY

Specifications

Description Dextrose Tryptone Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 33.3L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.