Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Dichloran Rose-Bengal Chloramphenicol (DRBC) Agar Base (Dehydrated)

Catalog No. OXCM0727B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0727B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0727B Supplier Thermo Scientific™ Supplier No. CM0727B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate and enumerate yeasts and molds associated with food spoilage with this selective medium.

Isolate and enumerate yeasts and molds associated with food spoilage with Thermo Scientific™ Oxoid™ Dichloran Rose-Bengal Chloramphenicol (DRBC) Agar Base (Dehydrated). DRBC Agar Base is specially designed to supress the growth of spreading mold and bacteria while supporting the growth of species that cannot be isolated on Rose Bengal Chloramphenicol Agar or Acidified Potato Dextrose Agar1.

Dichloran Rose-Bengal Chloramphenicol (DRBC) Agar Base is based on the formulation described by King et al.1,2, and is recommended as a selective medium for the isolation and enumeration of yeasts and molds that are of significance in food spoilage.

  • Easy-colony-counting: Rose-Bengal in the medium controls the size and height of mold colonies preventing overgrowth and making colony counting easier
  • Selective: appropriate for isolation of yeasts and molds from foods whose associated flora consists mainly of Gram-negative rod-shaped bacteria

DRBC Agar Base is a modification of Rose-Bengal Chloramphenicol Medium3 and differs as follows: pH is lowered to 5.6, the Rose-Bengal content is reduced by 50% and Dichloran is added.

The cumulative effect of these modifications is to further inhibit bacterial growth, inhibit spreading molds such as Rhizopus and Mucor and make the medium capable of supporting the growth of those species that cannot be isolated on Rose-Bengal Chloramphenicol Agar or acidified Potato Dextrose Agar1. The inhibition of spreading molds and the general restriction of colony size results in improved enumeration and detection of mycotoxigenic molds and other species of significance in food spoilage6.

Not all products are available for sale in all territories. Please inquire.


Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: NMKL standards.
Description DRBC Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 15.9L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.