Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ Endo Agar

Catalog No. oxcm0479b
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0479B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0479B Supplier Thermo Scientific™ Supplier No. CM0479B
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Isolate and differentiate Enterobacteriaceae from potable water, dairy products and foods with this differential medium.

Isolate and differentiate Enterobacteriaceae from potable water, dairy products and foods1, 2 with Thermo Scientific™ Oxoid™ Endo Agar (Dehydrated) when used with basic fuchsin BR0050. Originally designed for the isolation of typhoid bacillus3, Endo Agar is now mostly used for the differentiation of lactose fermenting and lactose non-fermenting intestinal organisms. Production of both acid and aldehyde by lactose fermenting organisms, such as Escherichia coli, gives rise to the characteristic red coloration of the colony and the surrounding medium.

Use Endo Agar for detection of coliform organisms in water, wastewater and dairy products for the presence of indicator groups such as coliforms. Besides safety, indicator groups provide a measure of food quality and spoilage potential. One of the most common groups of bacteria employed as indicator organisms by the food industry are the coliforms, which can be regarded as a subgroup within the Enterobacteriaceae.

  • Easy-to-read: lactose fermenting coliforms (e.g. Escherichia coli) give rise to deep red colonies which color the surrounding medium and possesses a golden metallic sheen. Non-lactose fermenters form colorless translucent colonies, against the pink to colorless medium.
  • Can be used for confirmation of presumptive test coliforms.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: APHA.
Description Endo Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 13.9L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.