Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Cronobacter sakazakii Isolation Agar (ISO) (Dehydrated)

Catalog No. CM1134B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CM1134B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CM1134B Supplier Thermo Scientific™ Supplier No. CM1134B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Detect and enumerate Chronobacter sakazakii in milk and milk samples as described in ISO TS/22964:2017 with this medium.

Detect and enumerate Cronobacter sakazakii in milk and milk samples as described in ISO TS/22964:2006 with Thermo Scientific™ Oxoid™ Cronobacter sakazakii Isolation Agar, when used in conjunction with Modified Lauryl Sulphate Broth (CM1133 and SR0247E). A chromogenic substrate present in the medium detects the α-glucosidase enzyme produced by Cronobacter sakazakii. Hydrolysis of the substrate results in blue-green colonies on this medium.

Cronobacter sakazakii is a Gram-negative rod-shaped bacterium that rarely causes disease in healthy adults. It has been implicated in outbreaks of disease in premature infants (neonates). Research suggests that neonates, or those infants who have other medical conditions, are more susceptible to this infection. Most reported cases of infection are severe, including sepsis (bacteria in the blood), meningitis, or necrotizing enterocolitis (severe intestinal infection).

Use Cronobacter sakazakii Isolation Agar for differentiation and enumeration of Cronobacter sakazakii from infant formula and other food samples.

  • Selective: Due to the presence of sodium desoxycholate and crystal violet the medium is selective against most Gram-positive organisms.
  • Easy-to-read: Cronobacter sakazakii produces blue-green colonies on this medium.

Cronobacter spp. have been isolated at low levels from powdered infant formulae, and the organisms’ high tolerance to desiccation provides a competitive advantage in the dry environments of milk powder factories, increasing the risk of product contamination1.

In response to concerns over this pathogen, a collaborative technical specification for the detection of Cronobacter in milk and milk products was jointly issued by ISO and IDF, as ISO/TS 22964:20062. This recommended pre-enrichment in BPW, selective enrichment in mLST/vancomycin broth and plating on Enterobacter sakazakii Isolation Agar. Positive colonies are confirmed on the basis of sugar utilisation, decarboxylase activity, as well as yellow pigmentation. The technical specification has since been superseded and among other changes the isolation medium has been replaced with Chromogenic Cronobacter Isolation Agar (CCI)3.

 

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description Enterobacter Sakazakii isolation Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 18.0L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.