Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Eosin Methylene Blue Agar (Modified) (Levine) (Dehydrated)
GSA_VA

Catalog No. oxcm0069b
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0069B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0069B Supplier Thermo Scientific™ Supplier No. CM0069B
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Detect, enumerate and differentiate the members of coliform group while conforming to the formula specified by APHA with this medium.

Detect, enumerate and differentiate the members of coliform group with Thermo Scientific™ Oxoid™ Eosin Methylene Blue Agar (Modified) Levine (Dehydrated). This versatile medium, modified by Levine1,2, is used for the differentiation of Escherichia coli and Enterobacteria aerogenes and also for rapid identification of Candida albicans. The medium is prepared to the formula specified by the APHA3,4,5,6 for the detection and differentiation of the coliform group of organisms7,8.

Use Eosin Methylene Blue Agar (Modified) Levine for isolation and differentiation of Enterobacterioceae.

  • Easy-to-read: due to Eosin–Y and methylene blue indicators Escherichia coli (purple colonies with green metallic sheen) are easy to differentiate from Enterobacteria aerogenes (purple mucoid colonies).
  • Recommended for the detection, enumeration, and differentiation of members of the coliform group by American Public Health Association.

Weld9,10 proposed the use of Eosin methylene blue agar (Modified) Levine, with added chlortetracycline hydrochloride for the rapid identification of Candida albicans in clinical materials. A positive identification of Candida albicans could be made after 24 to 48 hours incubation at 37°C in 10% carbon dioxide from feces, oral and vaginal secretions, and nail or skin scrapings.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Eosin-Methylene Blue Agar (Levine)
Form Powder
Product Type Agar
Quantity 500 g
Yield For 13.3L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.